Assay Details
| Gene Symbol: ptpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003425.2 | NP_001003425.2 | 4 - 5 | 406 | 99 | |
| GenBank mRNA | BC076243.1 | - | 4 - 5 | 370 | 99 | |
| EH505535.1 | - | 4 - 5 | 402 | 99 | ||
More Information
Set Membership: |
|

ptpa
Genomic Map