Assay Details
| Gene Symbol: cbx6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003768.1 | NP_001003768.1 | 1 - 2 | 102 | 70 | |
| XM_005161816.4 | XP_005161873.1 | 1 - 2 | 137 | 70 | ||
| XM_021469381.1 | XP_021325056.1 | 1 - 2 | 137 | 70 | ||
| GenBank mRNA | BC077116.1 | - | 1 - 2 | 102 | 70 | |
More Information
Set Membership: |
|

cbx6a
Genomic Map