Assay Details
| Gene Symbol: cbx6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003768.1 | NP_001003768.1 | 2 - 3 | 155 | 133 | |
| GenBank mRNA | BC077116.1 | - | 2 - 3 | 155 | 133 | |
More Information
Set Membership: |
|

cbx6a
Genomic Map