Assay Details
| Gene Symbol: akr1a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003783.2 | NP_001003783.1 | 5 - 6 | 591 | 134 | |
| GenBank mRNA | BC077140.1 | - | 5 - 6 | 595 | 134 | |
More Information
Set Membership: |
|

akr1a1a
Genomic Map