Assay Details
Gene Symbol: st3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004012.1 | NP_001004012.1 | 4 - 5 | 828 | 81 | |
XM_009293548.3 | XP_009291823.1 | 5 - 6 | 2593 | 81 | ||
XM_009293550.3 | XP_009291825.1 | 5 - 6 | 2352 | 81 | ||
GenBank mRNA | AJ705062.2 | - | 4 - 5 | 828 | 81 | |
AJ783741.1 | - | 4 - 5 | 828 | 81 | ||
BC162919.1 | - | 4 - 5 | 877 | 81 | ||
BC162930.1 | - | 4 - 5 | 877 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
