Assay Details
| Gene Symbol: st3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004012.1 | NP_001004012.1 | 5 - 6 | 947 | 60 | |
| XM_009293548.3 | XP_009291823.1 | 6 - 7 | 2712 | 60 | ||
| XM_009293550.3 | XP_009291825.1 | 6 - 7 | 2471 | 60 | ||
| GenBank mRNA | AJ705062.2 | - | 5 - 6 | 947 | 60 | |
| AJ783741.1 | - | 5 - 6 | 947 | 60 | ||
| BC162919.1 | - | 5 - 6 | 996 | 60 | ||
| BC162930.1 | - | 5 - 6 | 996 | 60 | ||
More Information
Set Membership: |
|

st3gal2
Genomic Map