Assay Details
| Gene Symbol: ndufs6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004651.1 | NP_001004651.1 | 3 - 4 | 338 | 65 | |
| GenBank mRNA | BC081528.1 | - | 3 - 4 | 338 | 65 | |
| BC164935.1 | - | 3 - 4 | 352 | 65 | ||
More Information
Set Membership: |
|

ndufs6
Genomic Map