Assay Details
| Gene Symbol: cox17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004652.3 | NP_001004652.1 | 3 - 4 | 340 | 89 | |
| GenBank mRNA | BC081527.1 | - | 3 - 4 | 306 | 89 | |
| EH490144.1 | - | 3 - 4 | 404 | 89 | ||
| EH495356.1 | - | 3 - 4 | 342 | 89 | ||
More Information
Set Membership: |
|

cox17
Genomic Map