Assay Details
| Gene Symbol: dhrs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006070.1 | NP_001006070.1 | 3 - 4 | 875 | 67 | |
| GenBank mRNA | BC083252.1 | - | 3 - 4 | 875 | 67 | |
| BC164070.1 | - | 3 - 4 | 493 | 67 | ||
More Information
Set Membership: |
|

dhrs3b
Genomic Map