Assay Details
| Gene Symbol: dhrs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006070.1 | NP_001006070.1 | 4 - 5 | 1110 | 88 | |
| GenBank mRNA | BC083252.1 | - | 4 - 5 | 1110 | 88 | |
| BC164070.1 | - | 4 - 5 | 728 | 88 | ||
More Information
Set Membership: |
|

dhrs3b
Genomic Map