Assay Details
| Gene Symbol: c1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007059.1 | NP_001007060.1 | 3 - 4 | 240 | 64 | |
| XM_021480568.1 | XP_021336243.1 | 3 - 4 | 249 | 64 | ||
| GenBank mRNA | BC083407.1 | - | 3 - 4 | 240 | 64 | |
More Information
Set Membership: |
|

c1d
Genomic Map