Assay Details
| Gene Symbol: mag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007062.2 | NP_001007063.2 | 1 - 2 | 83 | 60 | |
| NM_001313857.1 | NP_001300786.1 | 1 - 2 | 83 | 60 | ||
| XM_021481389.1 | XP_021337064.1 | 3 - 4 | 260 | 60 | ||
| XM_021481390.1 | XP_021337065.1 | 2 - 3 | 207 | 60 | ||
| XM_021481391.1 | XP_021337066.1 | 3 - 4 | 259 | 60 | ||
| XM_021481392.1 | XP_021337067.1 | 3 - 4 | 271 | 60 | ||
| XM_021481393.1 | XP_021337068.1 | 3 - 4 | 264 | 60 | ||
| XM_021481394.1 | XP_021337069.1 | 3 - 4 | 260 | 60 | ||
| XM_021481395.1 | XP_021337070.1 | 3 - 4 | 260 | 60 | ||
| GenBank mRNA | AJ628726.1 | - | 1 - 2 | 53 | 60 | |
| AJ628727.1 | - | 1 - 2 | 53 | 60 | ||
| CU458878.1 | - | 2 - 3 | 118 | 60 | ||
More Information
Set Membership: |
|

mag
Genomic Map