Assay Details
Gene Symbol: mnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009885.2 | NP_001009885.1 | 2 - 3 | 791 | 60 | |
GenBank mRNA | AY445044.1 | - | 2 - 3 | 659 | 60 | |
BC093243.1 | - | 2 - 3 | 791 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
