Assay Details
Gene Symbol: rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013345.1 | NP_001013363.1 | 3 - 4 | 233 | 77 | |
GenBank mRNA | BC090261.1 | - | 3 - 4 | 233 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
