Assay Details
| Gene Symbol: stk16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017630.1 | NP_001017630.1 | 7 - 8 | 919 | 73 | |
| GenBank mRNA | BC093283.1 | - | 7 - 8 | 919 | 73 | |
| BC164081.1 | - | 6 - 7 | 810 | 73 | ||
More Information
Set Membership: |
|

stk16
Genomic Map