Assay Details
Gene Symbol: fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017680.1 | NP_001017680.1 | 8 - 9 | 1233 | 69 | |
GenBank mRNA | BC093325.1 | - | 8 - 9 | 1233 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
