Assay Details
| Gene Symbol: lratd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017837.1 | NP_001017837.1 | 1 | 857 | 108 | |
| GenBank mRNA | BC092778.1 | - | 1 | 857 | 108 | |
More Information
Set Membership: |
|

lratd2b
Genomic Map