Assay Details
Gene Symbol: parp12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115086.1 | NP_001108558.1 | 4 - 5 | 933 | 121 | |
XM_009300122.3 | XP_009298397.1 | 3 - 4 | 543 | 121 | ||
GenBank mRNA | BC095326.1 | - | 4 - 5 | 903 | 121 | |
BC165181.1 | - | 4 - 5 | 846 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
