Assay Details
| Gene Symbol: amacr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020626.1 | NP_001018462.1 | 3 - 4 | 480 | 59 | |
| GenBank mRNA | BC095377.1 | - | 3 - 4 | 480 | 59 | |
More Information
Set Membership: |
|

amacr
Genomic Map