Assay Details
| Gene Symbol: sagb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033749.1 | NP_001028921.1 | 2 - 3 | 113 | 74 | |
| GenBank mRNA | BC080258.1 | - | 2 - 3 | 95 | 74 | |
| BC097245.1 | - | 2 - 3 | 113 | 74 | ||
More Information
Set Membership: |
|

sagb
Genomic Map