Assay Details
| Gene Symbol: sid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034182.2 | NP_001029354.2 | 438 | 64 | ||
| GenBank mRNA | BC134848.1 | - | 438 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sid4| Gene Symbol: sid4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034182.2 | NP_001029354.2 | 438 | 64 | ||
| GenBank mRNA | BC134848.1 | - | 438 | 64 | ||
Set Membership: |
|