Assay Details
| Gene Symbol: LOC572950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034992.1 | NP_001030164.1 | 3 - 4 | 440 | 70 | |
| Gene Symbol: jazf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021466680.1 | XP_021322355.1 | 3 - 4 | 700 | 70 | |
| GenBank mRNA | AY366304.1 | - | 3 - 4 | 440 | 70 | |
More Information
Set Membership: |
|

LOC572950
Genomic Map