Assay Details
| Gene Symbol: pla2g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037412.1 | NP_001032489.1 | 3 - 4 | 873 | 63 | |
| GenBank mRNA | BC108011.1 | - | 3 - 4 | 873 | 63 | |
More Information
Set Membership: |
|

pla2g3
Genomic Map