Assay Details
| Gene Symbol: stambpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037570.1 | NP_001032659.1 | 5 - 6 | 899 | 61 | |
| GenBank mRNA | BC110110.1 | - | 5 - 6 | 899 | 61 | |
More Information
Set Membership: |
|

stambpb
Genomic Map