Assay Details
Gene Symbol: tas1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039628.1 | NP_001034717.1 | 7 - 8 | 1983 | 87 | |
GenBank mRNA | AB200902.1 | - | 7 - 8 | 1983 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
