Assay Details
Gene Symbol: epas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039806.2 | NP_001034895.2 | 15 - 16 | 2645 | 65 | |
GenBank mRNA | BC163440.1 | - | 15 - 16 | 2422 | 65 | |
DQ375242.1 | - | 15 - 16 | 2596 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
