Assay Details
Gene Symbol: slc2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039808.1 | NP_001034897.1 | 4 - 5 | 743 | 83 | |
GenBank mRNA | DQ152824.1 | - | 4 - 5 | 743 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
