Assay Details
| Gene Symbol: casp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042695.1 | NP_001036160.1 | 2 - 3 | 298 | 87 | |
| XM_005157919.4 | XP_005157976.1 | 3 - 4 | 849 | 87 | ||
| XM_009292176.3 | XP_009290451.1 | 3 - 4 | 849 | 87 | ||
| XM_017351324.2 | XP_017206813.1 | 3 - 4 | 852 | 87 | ||
| XM_021466638.1 | XP_021322313.1 | 3 - 4 | 850 | 87 | ||
| GenBank mRNA | BC146717.1 | - | 3 - 4 | 745 | 87 | |
| BC163115.1 | - | 2 - 3 | 323 | 87 | ||
| BC163139.1 | - | 2 - 3 | 323 | 87 | ||
| DQ812119.1 | - | 2 - 3 | 298 | 87 | ||
More Information
Set Membership: |
|

casp2
Genomic Map