Assay Details
| Gene Symbol: epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043334.1 | NP_001036799.1 | 1 - 2 | 91 | 145 | |
| GenBank mRNA | BC163370.1 | - | 1 - 2 | 165 | 145 | |
| BC163373.1 | - | 1 - 2 | 166 | 145 | ||
| DQ273166.1 | - | 1 - 2 | 91 | 145 | ||
More Information
Set Membership: |
|

epor
Genomic Map