Assay Details
Gene Symbol: march2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044790.2 | NP_001038255.2 | 4 - 5 | 842 | 77 | |
GenBank mRNA | BC133146.1 | - | 4 - 5 | 842 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
