Assay Details
| Gene Symbol: scn4ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045065.1 | NP_001038530.1 | 10 - 11 | 1580 | 62 | |
| XM_005171585.4 | XP_005171642.1 | 11 - 12 | 1931 | 62 | ||
| GenBank mRNA | BC163558.1 | - | 10 - 11 | 1626 | 62 | |
| DQ149505.1 | - | 10 - 11 | 1580 | 62 | ||
More Information
Set Membership: |
|

scn4ab
Genomic Map