Assay Details
| Gene Symbol: scn4ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045065.1 | NP_001038530.1 | 5 - 6 | 704 | 64 | |
| XM_005171585.4 | XP_005171642.1 | 6 - 7 | 1055 | 64 | ||
| GenBank mRNA | BC163558.1 | - | 5 - 6 | 750 | 64 | |
| DQ149505.1 | - | 5 - 6 | 704 | 64 | ||
More Information
Set Membership: |
|

scn4ab
Genomic Map