Assay Details
| Gene Symbol: setd1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045134.2 | NP_001038599.2 | 10 - 11 | 2913 | 86 | |
| XM_005169219.4 | XP_005169276.1 | 9 - 10 | 3045 | 86 | ||
| XM_005169220.4 | XP_005169277.1 | 9 - 10 | 3045 | 86 | ||
| XM_009305666.3 | XP_009303941.1 | 9 - 10 | 3045 | 86 | ||
| XM_009305667.3 | XP_009303942.1 | 9 - 10 | 2860 | 86 | ||
| XM_009305669.3 | XP_009303944.1 | 9 - 10 | 3045 | 86 | ||
More Information
Set Membership: |
|

setd1ba
Genomic Map