Assay Details
| Gene Symbol: setd1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045134.2 | NP_001038599.2 | 16 - 17 | 5240 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

setd1ba| Gene Symbol: setd1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045134.2 | NP_001038599.2 | 16 - 17 | 5240 | 95 | |
Set Membership: |
|