Assay Details
Gene Symbol: gem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045849.1 | NP_001039314.1 | 1 - 2 | 572 | 84 | |
GenBank mRNA | BC122113.1 | - | 1 - 2 | 572 | 84 | |
KJ826437.1 | - | 1 - 2 | 313 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
