Assay Details
Gene Symbol: dctn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077212.1 | NP_001070680.1 | 6 - 7 | 855 | 81 | |
XM_021477458.1 | XP_021333133.1 | 8 - 9 | 917 | 81 | ||
GenBank mRNA | BC054565.1 | - | 6 - 7 | 856 | 81 | |
DQ141215.1 | - | 6 - 7 | 855 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
