Assay Details
| Gene Symbol: hsd17b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077328.1 | NP_001070796.1 | 5 - 6 | 702 | 57 | |
| GenBank mRNA | BC124783.1 | - | 5 - 6 | 702 | 57 | |
More Information
Set Membership: |
|

hsd17b7
Genomic Map