Assay Details
| Gene Symbol: retreg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080014.1 | NP_001073483.1 | 8 - 9 | 1003 | 59 | |
| GenBank mRNA | BC128884.1 | - | 8 - 9 | 1003 | 59 | |
More Information
Set Membership: |
|

retreg2
Genomic Map