Assay Details
Gene Symbol: rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082803.1 | NP_001076272.1 | 2 - 3 | 483 | 87 | |
GenBank mRNA | BC124310.1 | - | 2 - 3 | 510 | 87 | |
BC133897.1 | - | 2 - 3 | 483 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
