Assay Details
| Gene Symbol: pld6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089414.1 | NP_001082883.1 | 3 - 4 | 501 | 80 | |
| GenBank mRNA | BC122189.1 | - | 3 - 4 | 466 | 80 | |
| BC134035.1 | - | 3 - 4 | 501 | 80 | ||
More Information
Set Membership: |
|

pld6
Genomic Map