Assay Details
Gene Symbol: LOC558044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099989.1 | NP_001093459.1 | 20 - 21 | 3495 | 115 | |
XM_005165320.4 | XP_005165377.1 | 20 - 21 | 3886 | 115 | ||
XM_005165321.4 | XP_005165378.1 | 19 - 20 | 3884 | 115 | ||
XM_021476187.1 | XP_021331862.1 | 19 - 20 | 3387 | 115 | ||
XM_021476188.1 | XP_021331863.1 | 19 - 20 | 3387 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
