Assay Details
Gene Symbol: LOC558044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099989.1 | NP_001093459.1 | 29 - 30 | 5120 | 64 | |
XM_005165320.4 | XP_005165377.1 | 29 - 30 | 5511 | 64 | ||
XM_005165321.4 | XP_005165378.1 | 28 - 29 | 5509 | 64 | ||
XM_021476187.1 | XP_021331862.1 | 28 - 29 | 5012 | 64 | ||
XM_021476188.1 | XP_021331863.1 | 28 - 29 | 5012 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
