Assay Details
Gene Symbol: LOC558044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099989.1 | NP_001093459.1 | 39 - 40 | 6925 | 93 | |
XM_005165320.4 | XP_005165377.1 | 40 - 41 | 7418 | 93 | ||
XM_005165321.4 | XP_005165378.1 | 39 - 40 | 7416 | 93 | ||
XM_021476187.1 | XP_021331862.1 | 38 - 39 | 6817 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
