Assay Details
Gene Symbol: kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109869.2 | NP_001103339.2 | 4 - 5 | 939 | 97 | |
GenBank mRNA | BC153446.1 | - | 4 - 5 | 910 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
