Assay Details
| Gene Symbol: elk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130930.3 | NP_571005.2 | 3 - 4 | 1361 | 64 | |
| GenBank mRNA | AJ249170.1 | - | 3 - 4 | 1044 | 64 | |
More Information
Set Membership: |
|

elk4
Genomic Map