Assay Details
| Gene Symbol: sema3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131060.1 | NP_571135.1 | 13 - 14 | 1847 | 82 | |
| GenBank mRNA | AF086761.1 | - | 13 - 14 | 1847 | 82 | |
| BC162381.1 | - | 13 - 14 | 1560 | 82 | ||
| BC162398.1 | - | 13 - 14 | 1560 | 82 | ||
More Information
Set Membership: |
|

sema3aa
Genomic Map