Assay Details
| Gene Symbol: sema3aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131060.1 | NP_571135.1 | 3 - 4 | 693 | 58 | |
| GenBank mRNA | AF086761.1 | - | 3 - 4 | 693 | 58 | |
| BC162381.1 | - | 3 - 4 | 406 | 58 | ||
| BC162398.1 | - | 3 - 4 | 406 | 58 | ||
| CU458855.1 | - | 3 - 4 | 336 | 58 | ||
More Information
Set Membership: |
|

sema3aa
Genomic Map