Assay Details
Gene Symbol: rxrab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131153.1 | NP_571228.1 | 3 - 4 | 430 | 85 | |
XM_009301955.3 | XP_009300230.1 | 5 - 6 | 911 | 85 | ||
GenBank mRNA | U29894.1 | - | 3 - 4 | 430 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
