Assay Details
| Gene Symbol: ldhba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131247.1 | NP_571322.1 | 4 - 5 | 497 | 102 | |
| GenBank mRNA | AF067202.1 | - | 4 - 5 | 497 | 102 | |
| BC044190.1 | - | 4 - 5 | 513 | 102 | ||
| BC068981.1 | - | 4 - 5 | 486 | 102 | ||
More Information
Set Membership: |
|

ldhba
Genomic Map