Assay Details
Gene Symbol: ldhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131247.1 | NP_571322.1 | 5 - 6 | 670 | 76 | |
GenBank mRNA | AF067202.1 | - | 5 - 6 | 670 | 76 | |
BC044190.1 | - | 5 - 6 | 686 | 76 | ||
BC068981.1 | - | 5 - 6 | 659 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
